4-n-Nonylphenol, 98+%
4-n-Nonylphenol, 98+%
4-n-Nonylphenol, 98+%
4-n-Nonylphenol, 98+%
Thermo Scientific Chemicals
4-n-Nonylphenol, 98+%
CAS: 104-40-5 | C15H24O | 220.356 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15609.031 g
Catalog number ALFA15609.03
Price (MYR)
363.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
363.00
EA
Add to cart
Specifications
Chemical Name or Material4-n-Nonylphenol
CAS104-40-5
Health Hazard 1H302-H314-H335-H361fd
Health Hazard 2GHS H Statement
H314-H318-H361-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).
Solubility
Sparingly Soluble (0.020 g/L) at 25°C.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
RUO – Research Use Only
General References:
  1. Hyun Ju Lee; Soma Chattopadhyay; Eun-Yeung Gong; Ryun Sup Ahn; Keesook Lee. Antiandrogenic effects of bisphenol A and nonylphenol on the function of androgen receptor. Toxicological Sciences. 2003,75 (1), 40-46.
  2. Shigeru Kohtani; Shigeki Makino; Akihiko Kudo; Kunihiro Tokumura; Yasuhito Ishigaki;Tsukasa Matsunaga; Osamu Nikaido; Kazuichi Hayakawa; Ryoichi Nakagaki. Photocatalytic Degradation of 4-n-Nonylphenol under Irradiation from Solar Simulator: Comparison between BiVO4 and TiO2 Photocatalysts. Chemistry Letters. 2002, 31 (7), 660-661.