Benzo[b]furan-2-carboxylic acid, 98+%
Benzo[b]furan-2-carboxylic acid, 98+%
Benzo[b]furan-2-carboxylic acid, 98+%
Benzo[b]furan-2-carboxylic acid, 98+%
Benzo[b]furan-2-carboxylic acid, 98+%
CAS: 496-41-3 | C9H6O3 | 162.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15579.065 g
Catalog number ALFA15579.06
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzo[b]furan-2-carboxylic acid
CAS496-41-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c