Isopropylamine, 99+%
Isopropylamine, 99+%
Isopropylamine, 99+%
Isopropylamine, 99+%
Thermo Scientific Chemicals
Isopropylamine, 99+%
CAS: 75-31-0 | C3H9N | 59.112 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15044.AP500 mL
Catalog number ALFA15044.AP
Price (MYR)
308.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
308.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropylamine
CAS75-31-0
Health Hazard 1H224-H290-H301+H311+H331-H314-H335
Health Hazard 2GHS H Statement
H224-H315-H319-H335
Health Hazard 3P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
View more
Isopropylamine acts as a regulating agent for plastics and an additive in the petroleum industry. It is also useful as an intermediate for pharmaceuticals, coating materials, rubber chemicals and other organic products. It also finds application in binary chemical weapons in combination with isopropyl alcohol. Further, it is used as a solvent and to prepare other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropylamine acts as a regulating agent for plastics and an additive in the petroleum industry. It is also useful as an intermediate for pharmaceuticals, coating materials, rubber chemicals and other organic products. It also finds application in binary chemical weapons in combination with isopropyl alcohol. Further, it is used as a solvent and to prepare other chemicals.
Solubility
Miscible with water, chloroform, hexanes, acetone and ethanol.
Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and perchloryl fluoride.
RUO – Research Use Only
General References:
  1. Wen, X.; Zhang, H.; Xu, K.; Sun, J. H.; Ye, J.; Jin, S.; Wang, D. Crystal and molecular structures of twelve salts from isopropylamine and different organic acids. J. Mol. Struct. 2015, 1094, 46-60.
  2. Nikonov, A;. Y.; Sterkhova, I. V.; Lazareva, N. F. Reaction of N-[chloro(diorganyl)silyl]anilines with isopropanol and isopropylamine. Russ. J. Gen. Chem. 2015, 85 (8), 1866-1869.