3-Amino-1H-1,2,4-triazole, 96%
3-Amino-1H-1,2,4-triazole, 96%
3-Amino-1H-1,2,4-triazole, 96%
3-Amino-1H-1,2,4-triazole, 96%
Thermo Scientific Chemicals
3-Amino-1H-1,2,4-triazole, 96%
CAS: 61-82-5 | C2H4N4 | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1000 g
Catalog number ALFA14871.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Amino-1, 2, 4-triazole
CAS61-82-5
Health Hazard 1H302-H319-H350-H360-H373
Health Hazard 2GHS H Statement
H361-H373
Health Hazard 3P201-P202-P260-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
View more
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.
Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.
Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
RUO – Research Use Only
General References:
  1. Kailash C. Bhuyan.; Durga K. Bhuyan. Regulation of hydrogen peroxide in eye humors effect of 3-amino-1H-1,2,4-triazole on catalase and glutathione peroxidase of rabbit eye. Biochimica et Biophysica Acta (BBA) - General Subjects. 1977, 497, (3), 641-651.
  2. Grant R. Krow .; James T. Carey .; David E. Zacharias .; Edward E. Knaus. Reexamination of stereochemical issues concerning 2-phenyl-1,2-dihydropyridine-maleimide cycloadditions. J. Org. Chem. 1982, 47, (11), 1989-1993.