N-Benzylideneaniline, 99%
N-Benzylideneaniline, 99%
N-Benzylideneaniline, 99%
N-Benzylideneaniline, 99%
Thermo Scientific Chemicals
N-Benzylideneaniline, 99%
CAS: 538-51-2 | C13H11N | 181.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14746.1425 g
Catalog number ALFA14746.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Benzylideneaniline
CAS538-51-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzylideneaniline is an additional stain which is used in various immunoassays including flow cytometry, immunofluorescense, immunohistochemistry, and other applications.
Solubility
Soluble in water (partly), methanol, chloroform, alcohol, and acetic anhydride.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. K Weiss.; CH Warren.; G Wettermark. cis-trans isomerization about the carbon-nitrogen double bond. Structures of the isomers of N-benzylideneaniline. J. Am. Chem. Soc. 197193 (19), 4658-4663.
  2. J Bernstein.; YM Engel.; AT Hagler. An abinitio study of the conformational energetics of N-benzylideneaniline. J. Chem. Phys. 198175 (1), 2346-2348.