4-Pyridinemethanol, 99%
4-Pyridinemethanol, 99%
4-Pyridinemethanol, 99%
4-Pyridinemethanol, 99%
Thermo Scientific Chemicals
4-Pyridinemethanol, 99%
CAS: 586-95-8 | C6H7NO | 109.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14698.1425 g
Catalog number ALFA14698.14
Price (MYR)
477.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
477.00
EA
Add to cart
Specifications
Chemical Name or Material4-Pyridinemethanol
CAS586-95-8
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c