Tricine, 98+%
Tricine, 98+%
Tricine, 98+%
Thermo Scientific Chemicals
Tricine, 98+%
CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14695.1425 g
Catalog number ALFA14695.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTricine
CAS5704-04-1
Melting Point∼183°C (decomposition)
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Good's buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Good′s buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.
Solubility
Soluble in water.
Notes
pH range: 7.8-8.8Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Biological buffer, pKa = 8.15 at 20°: Biochemistry, 5, 467 (1966).
  2. Ying, Y.; Zhao, L.; Kong, L.; Kong, X.; Hua, Y.; Chen, Y. Solubilization of proteins in extracted oil bodies by SDS: A simple and efficient protein sample preparation method for Tricine-SDS-PAGE. Food Chem. 2015, 181, 179-185.
  3. Tapeinou, A.; Androutsou, M. E.; Kyrtata, K.; Vlamis-Gardikas, A.; Apostolopoulos, V.; Matsoukas, J.; Tselios, T. Conjugation of a peptide to mannan and its confirmation by tricine sodium dodecyl sulfate-polyacrylamide gel electrophoresis. Anal. Biochem. 2015, 485, 43-45.