4-Fluorobenzyl chloride, 99%
4-Fluorobenzyl chloride, 99%
4-Fluorobenzyl chloride, 99%
4-Fluorobenzyl chloride, 99%
Thermo Scientific Chemicals
4-Fluorobenzyl chloride, 99%
CAS: 352-11-4 | C7H6ClF | 144.573 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14686.22100 g
Catalog number ALFA14686.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Fluorobenzyl chloride
CAS352-11-4
Health Hazard 1H227-H314
Health Hazard 2GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Health Hazard 3P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c