4-Isopropylaniline, 99%
4-Isopropylaniline, 99%
4-Isopropylaniline, 99%
4-Isopropylaniline, 99%
Thermo Scientific Chemicals
4-Isopropylaniline, 99%
CAS: 99-88-7 | C9H13N | 135.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14646.1425 g
Catalog number ALFA14646.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Isopropylaniline
CAS99-88-7
Health Hazard 1H227-H302+H312+H332-H315-H319
Health Hazard 2GHS H Statement
H301-H311-H373-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
View more
4-Isopropylaniline was used in the hydroamination of phenylacetylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Isopropylaniline was used in the hydroamination of phenylacetylene.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong acids. Light sensitive
RUO – Research Use Only
S Breitenlechner.; M Fleck.; TE Müller. Solid catalysts on the basis of supported ionic liquids and their use in hydroamination reactions. Journal of Molecular Catalysis A: Chemical.2004214 (1), 175-179.