Glutaric acid, 99%
Glutaric acid, 99%
Glutaric acid, 99%
Thermo Scientific Chemicals
Glutaric acid, 99%
CAS: 110-94-1 | C5H8O4 | 132.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14595.22100 g
Catalog number ALFA14595.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlutaric acid
CAS110-94-1
Health Hazard 1H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
View more
Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.
Solubility
Soluble in water, alcohol, benzene and chloroform. Slightly soluble in petroleum ether.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Wang, Y.; Ma, J.; Zhou, Q.; Pang, S.; Zhang, Y. Hygroscopicity of Mixed Glycerol/Mg(NO3)2/Water Droplets Affected by the Interaction between Magnesium Ions and Glycerol Molecules. J. Phys. Chem. B. 2015,119 (17), 5558-5566.
  2. Wang, H.; Li, Z.; Yu, X.; Chen, R.; Xiulai Chen, X.; Liu, L. Green synthesis of (R)-3-TBDMSO glutaric acid methyl monoester using Novozym 435 in non-aqueous media. RSC Adv. 2015, 5 (92), 75160-75166.