2-Naphthol, 98+%
2-Naphthol, 98+%
2-Naphthol, 98+%
Thermo Scientific Chemicals
2-Naphthol, 98+%
CAS: 135-19-3 | C10H8O | 144.173 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14564.0B1000 g
Catalog number ALFA14564.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Naphthol
CAS135-19-3
Health Hazard 1H302+H332-H373
Health Hazard 2GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
View more
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
Solubility
Partly miscible in water (0.76 g/L).
Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. M. Panizza; P.A.Michaud, G.Cerisola; Ch. Comninellis. Anodic oxidation of 2-naphthol at boron-doped diamond electrodes. Journal of Electroanalytical Chemistry. 2001, 507, (1-2),206-214
  2. William R.Laws; Ludwig. Brand. Analysis of two-state excited-state reactions. The fluorescence decay of 2-naphthol. J. Phys. Chem. 1979, 83, (7),795-802