Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid
Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid
Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid
Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid
Thermo Scientific Chemicals
Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid
CAS: 37348-16-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
Catalog number ALFA14475.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrogen bromide
CAS37348-16-6
Health Hazard 1H226-H314-H318-H335
Health Hazard 2GHS H Statement
H314-H335-H227
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.