2-Undecanol, 98+%
2-Undecanol, 98+%
2-Undecanol, 98+%
Thermo Scientific Chemicals
2-Undecanol, 98+%
CAS: 1653-30-1 | C11H24O | 172.31 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14441.0910 g
Catalog number ALFA14441.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Undecanol
CAS1653-30-1
Health Hazard 1H315-H319
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
Melting Point1°C to 3°C
View more
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. István E. Markó, et al. Novel and Efficient Isomerization of Allylic Alcohols Promoted by a Tetrapropylammonium Perruthenate Catalyst.Angew. Chem. Int. Ed.,1999,38(13-14), 1960-1962.
  2. István E. Markó, et al. Efficient and Practical Catalytic Oxidation of Alcohols Using Molecular Oxygen.J. Org. Chem.,1988,66(22), 7576-7577.