2-(Trifluoromethyl)phenol, 98%
2-(Trifluoromethyl)phenol, 98%
2-(Trifluoromethyl)phenol, 98%
2-(Trifluoromethyl)phenol, 98%
Thermo Scientific Chemicals
2-(Trifluoromethyl)phenol, 98%
CAS: 444-30-4 | C7H5F3O | 162.111 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14231.065 g
Catalog number ALFA14231.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-(Trifluoromethyl)phenol
CAS444-30-4
Health Hazard 1H302+H312+H332-H318-H335
Health Hazard 2GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c