4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
Thermo Scientific Chemicals
4-(Trifluoromethyl)benzoyl chloride, 97%
CAS: 329-15-7 | C8H4ClF3O | 208.56 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14176.1425 g
Catalog number ALFA14176.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Trifluoromethyl)benzoyl chloride
CAS329-15-7
Health Hazard 1H227-H314
Health Hazard 2GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store under dry inert gas. Protect from humidity and water. Incompatible with water, bases, alcohol and oxidizing agents.
RUO – Research Use Only
General References:
  1. Viera Poláčková; Štefan Toma; Iveta Augustínová. Microwave-promoted cross-coupling of acid chlorides with arylboronic acids: a convenient method for preparing aromatic ketones. Tetrahedron.2006, 62 11675-11678.