1-Bromo-2,4-difluorobenzene, 98+%
1-Bromo-2,4-difluorobenzene, 98+%
1-Bromo-2,4-difluorobenzene, 98+%
1-Bromo-2,4-difluorobenzene, 98+%
Thermo Scientific Chemicals
1-Bromo-2,4-difluorobenzene, 98+%
CAS: 348-57-2 | C6H3BrF2 | 192.99 g/mol
Have Questions?
Catalog NumberQuantity
ALFA14098.1425 g
Catalog number ALFA14098.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromo-2,4-difluorobenzene
CAS348-57-2
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c