D-Isosorbide, 98%
D-Isosorbide, 98%
D-Isosorbide, 98%
Thermo Scientific Chemicals
D-Isosorbide, 98%
CAS: 652-67-5 | C6H10O4 | 146.142 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13989.1850 g
Catalog number ALFA13989.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Isosorbide
Melting Point56°C to 64°C
CAS652-67-5
Recommended StorageAmbient temperatures
RTECS NumberLZ4380000
View more
D-Isosorbide is used as a reagent in the preparation of detergents, agrochemicals, cleansers, cosmetics and vasodilators. It is also employed as a pharmaceutical intermediate for dimethyl ether and mono- and dinitrate derivatives, which is used as vasodilators and coronary disease therapeutics. It is used in the treatment of hydrocephalus and glaucoma. It finds application as a building block for polymers like polycarbonates and polyesters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in alcohols, water and ketones.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Parikh, K.; Mistry, B.; Jana, S.; Gajaria, T.; Gupta, S.; Devkar, R. V.; Kumar, S. Isosorbide spacer containing gemini surfactants: surface and biochemical study. Colloid. Polym. Sci. 2015, 293 (5), 1437-1446.
  2. Lavilla, C.; Munoz-Guerra, S. Sugar-based aromatic copolyesters: a comparative study regarding isosorbide and diacetalized alditols as sustainable comonomers. Green Chem. 2013, 15 (1), 144-151.