Copper(II) sulfate, anhydrous, 98%
Copper(II) sulfate, anhydrous, 98%
Copper(II) sulfate, anhydrous, 98%
Copper(II) sulfate, anhydrous, 98%
Copper(II) sulfate, anhydrous, 98%
Thermo Scientific Chemicals
Copper(II) sulfate, anhydrous, 98%
Copper(II) sulfate, 98%, CuO4S, CAS Number-7758-98-7, copper 2+ sulfate, copper ii sulfate, blue stone, copper monosulfate, copper sulphate, trinagle, copper sulfate, hylinec, cupric sulfate anhydrous, copper ii sulfate, anhydrous, 1000g, CHEBI:23414, 3.6, Hazard Class: 9; Packaging Group: III | CAS: 7758-98-7 | CuO4S | 159.602 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA13986.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper(II) sulfate
Melting Point>560°C
Name Noteanhydrous
CAS7758-98-7
Health Hazard 1H302-H315-H319
View more
Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling′s solution and Benedict′s solution to test for reducing sugars. Also, used to test protein in Biuret test.
Solubility
Soluble in water and methanol. Insoluble in ethanol.
Notes
Incompatible with strong bases, metals, alkali metals and powdered metals.
RUO – Research Use Only
General References:
  1. Dai, Q.; Zhu, X.; Abbas, S.; Karangwa, E.; Zhang, X.; Xia, S.; Feng, B.; Jia, C. Stable Nanoparticles Prepared by Heating Electrostatic Complexes of Whey Protein Isolate-Dextran Conjugate and Chondroitin Sulfate. J. Agric. Food Chem. 2015, 63 (16), 4179-4189.
  2. Lu, D.; Hossain, D.; Jia, Z.; Monteiro, M. J. One-Pot Orthogonal Copper-Catalyzed Synthesis and Self-Assembly of L-Lysine-Decorated Polymeric Dendrimers. Macromolecules 2015, 48 (6), 1688-1702.