Isopropyl 4-hydroxybenzoate, 98%
Isopropyl 4-hydroxybenzoate, 98%
Isopropyl 4-hydroxybenzoate, 98%
Isopropyl 4-hydroxybenzoate, 98%
Thermo Scientific Chemicals
Isopropyl 4-hydroxybenzoate, 98%
CAS: 4191-73-5 | C10H12O3 | 180.20 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13930.1850 g
Catalog number ALFA13930.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropyl 4-hydroxybenzoate
CAS4191-73-5
Health Hazard 1H315-H317-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
View more
It is used as a preservative in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a preservative in cosmetics.
Solubility
Soluble in Chloroform, Ethyl Acetate.
Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
RUO – Research Use Only
Anderson F. Alan. A thermochemical study of 1,10-decanediol. Journal of the American College of Toxicology. 1999, 14 (5), 53-57.