Glycylglycylglycine, 98+%
Glycylglycylglycine, 98+%
Glycylglycylglycine, 98+%
Glycylglycylglycine, 98+%
Thermo Scientific Chemicals
Glycylglycylglycine, 98+%
CAS: 556-33-2 | C6H11N3O4 | 189.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13778.065 g
Catalog number ALFA13778.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycylglycylglycine
Melting Point∼240°C (decomposition)
CAS556-33-2
Recommended StorageAmbient temperatures
EINECS Number209-122-0
View more
Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycylglycylglycine is used as a model peptide for studies of physicochemical parameters and molecular associations of small peptides. Triglycine is used as a copper chelator. It also acts as substrate for reproducible serum protein measurements by the biuret reaction1 and for the assay of aminotripeptidases
Solubility
Soluble in water (94.6 mg/ml at 20°C), methanol (50 mg/ml), and 80% acetic acid (50 mg/ml).
Notes
Store at -20°C. Protect from moisture. Avoid excessive heat and light.
RUO – Research Use Only
General References:
  1. MK Kumaran; ID Watson and GR Hedwig. Thermodynamic properties of peptide solutions. The relative partial molar enthalpies of aqueous solutions of glycylglycylglycine, glycylglycylalanine and alanylglycylglycine.Australian Journal of Chemistry.1983, 36 (9),1813 - 1820 .
  2. H. L. Yakel and E. W. Hughes. The unit-cell dimensions and space groups of two modifications of crystalline glycylglycylglycine.Acta Cryst.1952, 5 847-848 .