N-Benzylmethylamine, 97%
N-Benzylmethylamine, 97%
N-Benzylmethylamine, 97%
N-Benzylmethylamine, 97%
Thermo Scientific Chemicals
N-Benzylmethylamine, 97%
CAS: 103-67-3 | C8H11N | 121.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13746.AE100 mL
Catalog number ALFA13746.AE
Price (MYR)
281.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
281.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Benzylmethylamine
CAS103-67-3
Health Hazard 1H227-H301-H314-H335
Health Hazard 2GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
Solubility
Soluble in water (65 g/L at 20°C).
Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
RUO – Research Use Only
Harry F. Skinner. Methamphetamine synthesis via reductive alkylation hydrogenolysis of phenyl-2-propanone with N-benzylmethylamine. Forensic Science International.1993, 60 155-162.