4-Aminophenol, 98%
4-Aminophenol, 98%
4-Aminophenol, 98%
Thermo Scientific Chemicals
4-Aminophenol, 98%
CAS: 123-30-8 | C6H7NO | 109.128 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13581.1850 g
Catalog number ALFA13581.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Aminophenol
CAS123-30-8
Health Hazard 1H302+H332-H341
Health Hazard 2GHS H Statement
H341-H302-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P280-P301+P312-P304+P340-P308+P313-P312-P330-P501c
View more
4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminophenol is used as a dye for textiles, hair, furs and feathers. It is also used as a photographic developer. It finds application as a chemical intermediate in pharmaceuticals and dyes. Further, it acts as a final intermediate in the preparation of paracetamol.
Solubility
Soluble in acetonitrile, ethyl acetate, acetone, hot water, dimethylsulfoxide and alkalies. Slightly soluble in toluene, diethyl ether, ethanol, trifluoroacetic acid and cold water. Insoluble in benzene and chloroform.
Notes
Incompatible with acids, chloroformates and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Rahim, A.; Muhammad, N.; Nishan, U.; Khan, U. S.; Rehman, F.; Kubota, L. T.; Gushikem, Y. Copper phthalocyanine modified SiO2/C electrode as a biomimetic electrocatalyst for 4-aminophenol in the development of an amperometric sensor. RSC Adv. 2015, 5 (106), 87043-87050.
  2. Wang, J.; Shi, Z.; Jin, J.; Liu, Q.; Zhang, S. Determination of 4-aminophenol using a glassy carbon electrode modified with a three-dimensionally ordered macroporous film of polycysteine. Microchim. Acta 2015, 182 (3), 823-829.