Silicon carbide powder, superfine, 600 grit
Silicon carbide powder, superfine, 600 grit
Silicon carbide powder, superfine, 600 grit
Silicon carbide powder, superfine, 600 grit
Thermo Scientific Chemicals
Silicon carbide powder, superfine, 600 grit
CAS: 409-21-2 | CSi | 40.10 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13561.36500 g
Catalog number ALFA13561.36
Price (MYR)
651.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
651.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilicon carbide powder
Name Notesuperfine, 600 grit
CAS409-21-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only