2,4-Dihydroxybenzoic acid, 97%
2,4-Dihydroxybenzoic acid, 97%
2,4-Dihydroxybenzoic acid, 97%
2,4-Dihydroxybenzoic acid, 97%
Thermo Scientific Chemicals
2,4-Dihydroxybenzoic acid, 97%
CAS: 89-86-1 | C7H6O4 | 154.121 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13545.36500 g
Catalog number ALFA13545.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Dihydroxybenzoic acid
CAS89-86-1
Health Hazard 1H315-H319-H335-H500
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.
Solubility
Soluble in water(70g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Florence Benoit-Marquie.; Edith Puech-Costes.; AndreM. Braun.; Esther Oliveros.; Marie-Therese Maurette. Photocatalytic degradation of 2,4-dihydroxybenzoic acid in water: efficiency optimization and mechanistic investigations. Journal of Photochemistry and Photobiology A: Chemistry. 1997, 108, (1), 65-71.
  2. De Lima LeiteR.H.; Cognet, P.; Wilhelm, A.M.; Delmas, H. Anodic oxidation of 2,4-dihydroxybenzoic acid for wastewater treatment: study of ultrasound activation. Chemical Engineering Science. 2002, 57, (5), 767-778.