p-Cresol, 99%
p-Cresol, 99%
p-Cresol, 99%
Thermo Scientific Chemicals
p-Cresol, 99%
CAS: 106-44-5 | C7H8O | 108.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13531.30250 g
Catalog number ALFA13531.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Materialp-Cresol
CAS106-44-5
Health Hazard 1H301+H311-H314-H335
Health Hazard 2GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.
Solubility
Soluble in ethanol, diethyl ether and acetone. Slightly soluble in water.
Notes
Air and light sensitive. Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only
General References:
  1. Zhang, L.; Jiang, L. Y.; Zheng, K.; Duan, H.; Li, M.; Cui, Y. C. Enzymatic Synthesis of p-Cresol Oligomers and Evaluation of their Free Radical Scavenging Activity. Aust. J. Chem. 2014, 68 (2), 282-287.
  2. Fitzgerald, S.; Kolar, P.; Classen, J.; Boyette, M.; Das, L. Swine manure char as an adsorbent for mitigation of p-cresol. Environ. Prog. Sustainable Energy 2015, 34 (1), 125-131.