L-Aspartic acid, 98+%
L-Aspartic acid, 98+%
L-Aspartic acid, 98+%
L-Aspartic acid, 98+%
Thermo Scientific Chemicals
L-Aspartic acid, 98+%
CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA13520.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Aspartic acid
Melting Point>300°C (decomposition)
CAS56-84-8
Recommended StorageAmbient temperatures
RTECS NumberCI9098500
View more
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
Solubility
Soluble in water(4.5g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Flaig, R.; Koritsanszky, T.; Zobel, D.; Luger, P. Topological Analysis of the Experimental Electron Densities of Amino Acids. 1. d,l-Aspartic Acid at 20 K. J. Am. Chem. Soc. 1998, 120, (10), 2227-2238.
  2. Xiao-Lin Wang.; Ai-Ling Ying.; Wei-Ning Wang. Nanofiltration of l-phenylalanine and l-aspartic acid aqueous solutions. Journal of Membrane Science. 2002, 196, (1), 59-67.
  3. Readily forms ß-lactam derivatives whichare valuable intermediates for the synthesis of a variety of products, including thienamycin: J. Am. Chem. Soc., 102, 6163 (1980); Tetrahedron Lett., 23, 2293 (1982).