(+/-)-4-Methyl-2-pentanol, 99%
(+/-)-4-Methyl-2-pentanol, 99%
(+/-)-4-Methyl-2-pentanol, 99%
(+/-)-4-Methyl-2-pentanol, 99%
Thermo Scientific Chemicals
(+/-)-4-Methyl-2-pentanol, 99%
CAS: 108-11-2 | C6H14O | 102.177 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13435.AP500 mL
Catalog number ALFA13435.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-4-Methyl-2-pentanol
CAS108-11-2
Health Hazard 1H226-H335
Health Hazard 2GHS H Statement
H226-H335
Flammable liquid and vapour.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
View more
(±)-4-Methyl-2-pentanol is employed as a solvent in organic synthesis, dyes and stains, natural resins and waxes. It is an additive to surface coatings. It finds applications in the manufacture of lube oil additives, zinc dialkyldithiophosphate, brake fluids. It is the precursor to some plasticizers and frother in mineral flotation. It acts as a corrosion inhibitor and also utilized in the characterization of volatile fraction of mono varietal wines and tar sand mining processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(^+)-4-Methyl-2-pentanol is employed as a solvent in organic synthesis, dyes and stains, natural resins and waxes. It is an additive to surface coatings. It finds applications in the manufacture of lube oil additives, zinc dialkyldithiophosphate, brake fluids. It is the precursor to some plasticizers and frother in mineral flotation. It acts as a corrosion inhibitor and also utilized in the characterization of volatile fraction of mono varietal wines and tar sand mining processes.
Solubility
Miscible with ethanol and diethyl ether. Slightly miscible with water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and acid chlorides.
RUO – Research Use Only
General References:
  1. Mizobuchi, M.; Nagayama, K. Reduced pressure gas phase bioreactor as a tool for stereoselective reduction catalyzed by alcohol dehydrogenase from Parvibaculum lavamentivorans. Biochem. Eng. J. 2015, 93, 11-16.
  2. Bournival, G.; Souza, L. O.; Ata, S.; Wanless, E. J. Effect of alcohol frothing agents on the coalescence of bubbles coated with hydrophobized silica particles. Chem. Eng. Sci. 2015, 131, 1-11.