Pyrogallol, 98+%
Pyrogallol, 98+%
Pyrogallol, 98+%
Thermo Scientific Chemicals
Pyrogallol, 98+%
CAS: 87-66-1 | C6H6O3 | 126.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13405.22100 g
Catalog number ALFA13405.22
Price (MYR)
427.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
427.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyrogallol
CAS87-66-1
Health Hazard 1H301-H312+H332-H315-H319-H341
Health Hazard 2GHS H Statement
H341-H302-H312-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
View more
Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal-complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal-complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.
Solubility
Soluble in ethanol, water and ether. Slightly soluble in chloroform.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, alkalis, metal oxides, ammonia, iodine, lime water, potassium permanganate and strong bases.
RUO – Research Use Only
General References:
  1. Can, M.; Bulut, E.; Ozacar, M. Reduction of palladium onto pyrogallol-derived nano-resin and its mechanism. Chem. Eng. J. 2015, 275, 322-330.
  2. Guralnik, V.; Avram, L.; Cohen, Y. Unique Organization of Solvent Molecules Within the Hexameric Capsules of Pyrogallol [4] arene in Solution. Org. Lett. 2014, 16 (21), 5592-5595.