Resorcinol, 99%
Resorcinol, 99%
Resorcinol, 99%
Thermo Scientific Chemicals
Resorcinol, 99%
CAS: 108-46-3 | C6H6O2 | 110.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA13080.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialResorcinol
CAS108-46-3
Health Hazard 1H301-H315-H319-H500
Health Hazard 2GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
View more
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.
Solubility
Soluble in water, alcohol, ether and glycerol.
Notes
Stable. May discolor on exposure to air or light.
RUO – Research Use Only
General References:
  1. Zhang, J. Z.; Fischer, C. J. A simplified resorcinol method for direct spectrophotometric determination of nitrate in seawater. Marine Chemistry 2006, 99 (1-4), 220-226.
  2. Schwandt, N. W.; Gound, T. G. Resorcinol-Formaldehyde Resin Russian Red Endodontic Therapy. J. Endod. 2003, 29 (7), 435-437.