1,1,2,2-Tetrabromoethane, 97%
1,1,2,2-Tetrabromoethane, 97%
1,1,2,2-Tetrabromoethane, 97%
1,1,2,2-Tetrabromoethane, 97%
Thermo Scientific Chemicals
1,1,2,2-Tetrabromoethane, 97%
CAS: 79-27-6 | C2H2Br4 | 345.65 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12943.AU1000 mL
Catalog number ALFA12943.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or Material1,1,2,2-Tetrabromoethane
CAS79-27-6
Health Hazard 1H319-H330
Health Hazard 2GHS H Statement
H330-H319
Fatal if inhaled.
Causes serious eye irritation.
Health Hazard 3P260-P271-P284-P304+P340-P305+P351+P338-P310-P501c
View more
1,1,2,2-Tetrabromoethane is useful for the separation of minerals from supporting rock. It is used as a solvent for fats, greases, oils and waxes. It acts as a mercury substitute in the production of gauges and balancing equipment. It finds application as a floating agent used in processing and separation of mineral oils. Further, it is used as refractive index liquid in microscopy. In addition to this, it is also employed as a flame-retardant additive for polystyrenes, polyurethanes and polyolefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,2,2-Tetrabromoethane is useful for the separation of minerals from supporting rock. It is used as a solvent for fats, greases, oils and waxes. It acts as a mercury substitute in the production of gauges and balancing equipment. It finds application as a floating agent used in processing and separation of mineral oils. Further, it is used as refractive index liquid in microscopy. In addition to this, it is also employed as a flame-retardant additive for polystyrenes, polyurethanes and polyolefins.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, aluminum, magnesium, alkali metals and zinc.
RUO – Research Use Only
General References:
  1. Petrov, A. R.; Derheim, A.; Oetzel, J.; Leibold, M.; Bruhn, C.; Scheerer, S.; Oßwald, S.; Winter, R. F.; Siemeling, U. A Stable Planar-Chiral N-Heterocyclic Carbene with a 1,1'-Ferrocenediyl Backbone. Inorg. Chem. 2015, 54 (13), 6657-6670.
  2. Afreh, I.; Wujcik, E. K.; Blasdel, N.; Sauer, B.; Kaya, S.; Duirk, S.; Monty, C. N. Detection of halogenated organics by their inhibitory action in a catalytic reaction between dimethyl acetylenedicarboxylate and 2-methyl-4-nitroaniline. J. Anal. Chem. 2015, 70 (7), 825-830.