2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
Thermo Scientific Chemicals
2-Acetylpyridine, 98%
CAS: 1122-62-9 | C7H7NO | 121.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA12688.22
Price (MYR)
744.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
744.00
EA
Add to cart
Specifications
Chemical Name or Material2-Acetylpyridine
CAS1122-62-9
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
2-Acetylpyridine used as flavoring agent. It is also used in application for food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Acetylpyridine used as flavoring agent. It is also used in application for food additive.
Solubility
Soluble in water (18.2 g/100g @ 25C). Soluble in and acetate. Slightly soluble in carbon tetrachloride.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Daniel L. Klayman; Joseph F. Bartosevich; T. Scott Griffin; Carl J. Mason; John P. Scovill. 2-Acetylpyridine thiosemicarbazones. 1. A new class of potential antimalarial agents. J. Med. Chem. 1979, 22(7), 145-150.
  2. John P. Scovill; Daniel L. Klayman; Charles F. Franchino. 2-Acetylpyridine thiosemicarbazones. 4. Complexes with transition metals as antimalarial and antileukemic agents. J. Med. Chem. 1982, 2510), 1261-1264.