Polytetrafluoroethylene
Polytetrafluoroethylene
Polytetrafluoroethylene
Polytetrafluoroethylene
Thermo Scientific Chemicals
Polytetrafluoroethylene
CAS: 9002-84-0 | (C2F4)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA12613.30
Price (MYR)
1,579.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
1,579.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolytetrafluoroethylene
CAS9002-84-0
Recommended StorageAmbient temperatures
RTECS NumberKX4025000
TSCAYes
View more
PTFE may be used as a non stick coating. It can be used as engineering plastics. It is employed as lining reaction vessels, gaskets and pump packing, electrical insulator for high frequency applications.