Selenium, powder, 99+%
Selenium, powder, 99+%
Actual product may vary
Thermo Scientific Chemicals
Selenium, powder, 99+%
Have Questions?
Catalog NumberQuantity
ALFA12592.30250 g
Catalog number ALFA12592.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point690°C
Melting Point224°C to 225°C
Quantity250 g
Percent Purity≥99%
Unit SizeEA