2-Methyl-4(5)-nitroimidazole, 99%
2-Methyl-4(5)-nitroimidazole, 99%
2-Methyl-4(5)-nitroimidazole, 99%
2-Methyl-4(5)-nitroimidazole, 99%
Thermo Scientific Chemicals
2-Methyl-4(5)-nitroimidazole, 99%
CAS: 696-23-1 | C4H5N3O2 | 127.10 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12569.1850 g
Catalog number ALFA12569.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-4(5)-nitroimidazole
CAS696-23-1
Health Hazard 1H302
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c