trans-Cinnamoyl chloride, 97%
trans-Cinnamoyl chloride, 97%
trans-Cinnamoyl chloride, 97%
trans-Cinnamoyl chloride, 97%
Thermo Scientific Chemicals
trans-Cinnamoyl chloride, 97%
CAS: 17082-09-6 | C9H7ClO | 166.604 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12016.22100 g
Catalog number ALFA12016.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-Cinnamoyl chloride
CAS17082-09-6
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The derivative of this compound, trans-Cinnamoylimidazole is involved in the synthesis of trans-Cinnamoyl-Papain.
Solubility
Reacts with water.
Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases. Moisture sensitive.
RUO – Research Use Only
General References:
  1. Lewis J. Brubacher.; Myron L. Bender. The Preparation and Properties of trans-Cinnamoyl-Papain. J. Am. Chem. Soc. 1966, 88 (24),5871-5880.
  2. Stefan Latza.; Dietmar Gansser.; Ralf G. Berger. Identification and Accumulation of 1-O-trans-Cinnamoyl-β-d-glucopyranose in Developing Strawberry Fruit (Fragaria ananassa Duch. Cv. Kent). J. Agric. Food Chem. 1996, 44 (6),1367-1370.