Lithium L-lactate, 97%
Lithium L-lactate, 97%
Lithium L-lactate, 97%
Lithium L-lactate, 97%
Thermo Scientific Chemicals
Lithium L-lactate, 97%
CAS: 27848-80-2 | C3H5LiO3 | 96.01 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11818.1850 g
Catalog number ALFA11818.18
Price (MYR)
535.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
535.00
EA
Add to cart
Specifications
Chemical Name or MaterialLithium L-lactate
CAS27848-80-2
Melting Point253°C to 255°C
Recommended StorageAmbient temperatures
RTECS NumberOJ6311000
View more
Useful chiral synthon, building block for depsipeptides