4-Methoxy-2-nitroaniline, 99%
4-Methoxy-2-nitroaniline, 99%
4-Methoxy-2-nitroaniline, 99%
4-Methoxy-2-nitroaniline, 99%
Thermo Scientific Chemicals
4-Methoxy-2-nitroaniline, 99%
CAS: 96-96-8 | C7H8N2O3 | 168.152 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11673.22100 g
Catalog number ALFA11673.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Methoxy-2-nitroaniline
CAS96-96-8
Health Hazard 1H300+H310+H330-H373
Health Hazard 2GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
View more
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.
Solubility
Slightly soluble in water. Soluble in hot methanol almost transparency.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. C. Ravikumara; and I. Hubert Joe. Electronic absorption and vibrational spectra and nonlinear optical properties of 4-methoxy-2-nitroaniline. Phys. Chem. Chem. Phys. 2010, 12 (32), 9452-9460.
  2. R.H. Contreras; D.G. De Kowalewski; J.C. Facelli .The NMR analysis of the methoxy-group conformation in 4-methoxy-2-nitroaniline. Journal of Molecular Structure. 1982, 81 (1-2), 147-149.