Aluminum acetate, basic hydrate
Aluminum acetate, basic hydrate
Aluminum acetate, basic hydrate
Aluminum acetate, basic hydrate
Thermo Scientific Chemicals
Aluminum acetate, basic hydrate
CAS: 142-03-0 | C4H7AlO5 | 162.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA11620.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum acetate
Name NoteBasic hydrate
CAS142-03-0
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.
Solubility
Insoluble in water
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Al-Ghamdi, A. A.; Al-Hartomy, O. A.; El Okr, M.; Nawar, A. M.; El-Gazzar, S.; El-Tantawy, F.; Yakuphanoglu, F. Semiconducting properties of Al doped ZnO thin films. Spectrochim. Acta, Part A 2014, 131, 512-517.
  2. Shin, H. U.; Stefaniak, A. B.; Stojilovic, N.; Chase, G. G. Comparative dissolution of electrospun Al2O3 nanofibres in artificial human lung fluids. Environ. Sci.: Nano 2015, 2 (3), 251-261.