1-Decyne, 98%
1-Decyne, 98%
1-Decyne, 98%
Thermo Scientific Chemicals
1-Decyne, 98%
CAS: 764-93-2 | C10H18 | 138.254 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11171.065 g
Catalog number ALFA11171.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Decyne
CAS764-93-2
Health Hazard 1H226-H315-H318
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
Melting Point-44°C
View more
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.
Solubility
Not miscible in water.
Notes
Incompatible with strong oxidizing agents. Store at 0-6°C.
RUO – Research Use Only
General References:
  1. Lucy L; Fan; Jiunn- Yann Tang; Alan Wohlman. Investigation of 1-decyne formation in cottonseed oil fried foods. Journal of the American Oil Chemists' Society. 1983, 60 (5), 1115-1119.
  2. For details of hydroalumination (DIBAH) and subsequent palladium(0)-catalyzed coupling with (Z)-1-hexenyl iodide, see: Org. Synth. Coll., 8, 295 (1993).