4-(4-Methoxyphenyl)-2-butanone, 99%
4-(4-Methoxyphenyl)-2-butanone, 99%
4-(4-Methoxyphenyl)-2-butanone, 99%
4-(4-Methoxyphenyl)-2-butanone, 99%
Thermo Scientific Chemicals
4-(4-Methoxyphenyl)-2-butanone, 99%
CAS: 104-20-1 | C11H14O2 | 178.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFA11011.1850 g
Catalog number ALFA11011.18
Price (MYR)
0.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
0.00
EA
Add to cart
Specifications
Chemical Name or Material4- (4-Methoxyphenyl)-2-butanone
CAS104-20-1
Melting Point7°C to 9°C
Recommended StorageAmbient temperatures
Density1.046
View more
4-(4-Methoxyphenyl)-2-butanone is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(4-Methoxyphenyl)-2-butanone is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Katsuhiko Nakamichi; Takeji Shibatani; Yoko Yamamoto; Tadashi Sato. Asymmetric amination of 4-methoxyphenylacetone and its related compounds with microorganisms. Applied Microbiology and Biotechnology. 1990, 33,(6), 637-640.
  2. Maria J.Climent; Avelino Corma; Sara Iborra; Maria Mifsud; Alexandra Velty. New one-pot multistep process with multifunctional catalysts: decreasing the E factor in the synthesis of fine chemicals. Green Chem. 2010, 12,(1), 99-107.