Propionyl chloride, 98%
Propionyl chloride, 98%
Propionyl chloride, 98%
Propionyl chloride, 98%
Thermo Scientific Chemicals
Propionyl chloride, 98%
CAS: 79-03-8 | C3H5ClO | 92.522 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10940.22100 g
Catalog number ALFA10940.22
Price (MYR)
285.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
285.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropionyl chloride
CAS79-03-8
Health Hazard 1H225-H302-H314-H331-H335
Health Hazard 2GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
Propionyl chloride is used as an intermediate in the production of agrochemicals and pharmaceuticals. It serves as an intermediate for dyes, textile auxiliaries and peroxide compounds. It acts as a crop protecting agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propionyl chloride is used as an intermediate in the production of agrochemicals and pharmaceuticals. It serves as an intermediate for dyes, textile auxiliaries and peroxide compounds. It acts as a crop protecting agent.
Notes
Moisture sensitive. Incompatible with alcohols, oxidizing agents, strong bases and water.
RUO – Research Use Only
General References:
  1. Snead, D. R.; Jamison, T. F. A Three-Minute Synthesis and Purification of Ibuprofen: Pushing the Limits of Continuous-Flow Processing. Angew. Chem. 2015, 127 (3), 997-1001.
  2. Lu, T.; Lu, C.; Nie, J.; Chen, Z.; Yang, G. Non-Cross-Linked Polystyrene Supported Cyclosulfamide as a New Chiral Auxiliary for Asymmetric Aldol Reactions. Curr. Org. Synth. 2015, 12 (1), 61-66.