Benzophenone, 99%
Benzophenone, 99%
Benzophenone, 99%
Thermo Scientific Chemicals
Benzophenone, 99%
CAS: 119-61-9 | C13H10O | 182.222 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA10739.30
Price (MYR)
374.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
374.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzophenone
CAS119-61-9
Health Hazard 1H351-H373-H500
Health Hazard 3P201-P202-P260-P281-P308+P313-P501c
Melting Point47°C to 50°C
View more
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzophenone is widely employed as a photo initiator in UV-curing applications such as inks, imaging and clear coatings in the printing industry. It acts as a UV blocker to prevent photo-degradation of the packaging polymers or its contents. It protects some products such as perfumes and soaps from ultraviolet light. It serves to manufacturers to package the product in clear glass or plastic.
Solubility
Soluble in ethanol and diethyl ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. For a review of the use of benzophenone as a triplet-sensitizer in photochemistry, see: Synthesis, 249 (1981).
  2. Chung, W. H.; Tzing, S. H.; Ding, W. H. Optimization of dispersive micro solid-phase extraction for the rapid determination of benzophenone-type ultraviolet absorbers in aqueous samples. J. Chromatogr. A 2015, 1411, 17-22.
  3. Dias, A. N.; Silva, A. C.; Simao, V.; Merib, J.; Carasek, E. A novel approach to bar adsorptive microextraction: Cork as extractor phase for determination of benzophenone, triclocarban and parabens in aqueous samples. Anal. Chim. Acta 2015, 888, 59-66.