3,5-Dimethylaniline, 97+%
3,5-Dimethylaniline, 97+%
3,5-Dimethylaniline, 97+%
3,5-Dimethylaniline, 97+%
Thermo Scientific Chemicals
3,5-Dimethylaniline, 97+%
CAS: 108-69-0 | C8H11N | 121.183 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10570.22100 g
Catalog number ALFA10570.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3,5-Dimethylaniline
Melting Point9°C to 11°C
CAS108-69-0
Health Hazard 1H227-H301+H311+H331-H373
Health Hazard 2GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
View more
3,5-Dimethylaniline is the reagent commonly useful in the manufacture of dyes. It undergoes polymerization in the presence of cerium(IV) sulfate as an oxidant. 3,5-Dimethylaniline was used in the synthesis of chiral packing materials for high performance liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dimethylaniline is the reagent commonly useful in the manufacture of dyes. It undergoes polymerization in the presence of cerium(IV) sulfate as an oxidant. 3,5-Dimethylaniline was used in the synthesis of chiral packing materials for high performance liquid chromatography.
Solubility
Soluble in water. <0.1 g/100 mL at 19°C
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Air and light sensitive. Incompatible to acids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates, halogens.
RUO – Research Use Only
General References:
  1. L Cui.; HL Sun.; JS Wishnok. Identification of adducts formed by reaction of N-acetoxy-3, 5-dimethylaniline with DNA. Chem. Res. Toxicol.2007 20, (11) , 1730-1736.
  2. P Snauwaert.; R Lazzaron.; J Riga.; JJ Verbist. Electronic structure and properties of dimethyl-aniline polymers. Synthetic Metals.1987 21, (1-3) , 181-187.