Indole-3-acetic acid, 98+%
Indole-3-acetic acid, 98+%
Indole-3-acetic acid, 98+%
Indole-3-acetic acid, 98+%
Thermo Scientific Chemicals
Indole-3-acetic acid, 98+%
CAS: 87-51-4 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA10556.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIndole-3-acetic acid
CAS87-51-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.
Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Cheryl L. Patten; Bernard R. Glick. Bacterial biosynthesis of indole-3-acetic acid. Can. J. Microbiol. 1996, 42, (3), 207-220.
  2. J. RIGAUD, A. PUPPO. Indole-3-acetic Acid Catabolism by Soybean Bacteroids. Microbiology. 1975, 88, 223-228.