Valerophenone, 99%
Valerophenone, 99%
Valerophenone, 99%
Thermo Scientific Chemicals
Valerophenone, 99%
CAS: 1009-14-9 | C11H14O | 162.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA10525.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialValerophenone
CAS1009-14-9
Melting Point-9°C
Recommended StorageAmbient temperatures
Density0.978
View more
Valerophenone is used as intermediates of liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Valerophenone is used as intermediates of liquid crystals.
Solubility
Insoluble in water.
Notes
 Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
RUO – Research Use Only
General References:
  1. R.G.Zepp; M.M.Gumz; W.L.Miller; H.Gao. Photoreaction of Valerophenone in Aqueous Solution. J. Phys. Chem. A. 1998, 102, (28),5716-5723
  2. P.Novak; J.Matousek; J.Briza. Valerophenone synthase-like chalcone synthase homologues in Humulus lupulus. Biologia Plantarum. 2003, 46, (3),375-381