Gadolinium(III) acetate hydrate, 99.9% (REO)
Gadolinium(III) acetate hydrate, 99.9% (REO)
Gadolinium(III) acetate hydrate, 99.9% (REO)
Gadolinium(III) acetate hydrate, 99.9% (REO)
Gadolinium(III) acetate hydrate, 99.9% (REO)
Thermo Scientific Chemicals
Gadolinium(III) acetate hydrate, 99.9% (REO)
CAS: 100587-93-7 | C6H9GdO6 | 334.38 g/mol
Have Questions?
Catalog NumberQuantity
ALF089158.22100 g
Catalog number ALF089158.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point(decomposition)
Chemical Name or MaterialGadolinium(III) acetate hydrate
CAS100587-93-7
ColorWhite
Health Hazard 1H315-H319-H335
View more
Gadolinium(III) acetate hydrate is used as a laboratory reagent, catalysts, optical glasses, structural ceramics, electrical components and photo-optical material.