Sodium tetraethylborate, 98%
Sodium tetraethylborate, 98%
Sodium tetraethylborate, 98%
Sodium tetraethylborate, 98%
Thermo Scientific Chemicals
Sodium tetraethylborate, 98%
CAS: 15523-24-7 | C8H20BNa | 150.048 g/mol
Have Questions?
Catalog NumberQuantity
ALF089137.031 g
Catalog number ALF089137.03
Price (MYR)
1,101.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,101.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium tetraethylborate
CAS15523-24-7
Health Hazard 1H250-H261
Health Hazard 2GHS H Statement
H250-H260-H252
Health Hazard 3P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c
View more
Sodium tetraethylborate is used as a derivatizing agent for the detection of organotin, lead and mercury compounds by using gas chromatography coupled with various sensitive and selective detection methods, such as atomic absorption spectrometry (AAS), atomic emission spectrometry (AES), pulsed-flame photometric detection (PFPD), and mass spectrometry (MS). It is also used as an alkylating agent.