Silver, AAS standard solution, Specpure™ Ag 1000μg/mL, Thermo Scientific Chemicals
Silver, AAS standard solution, Specpure™ Ag 1000μg/mL, Thermo Scientific Chemicals
Silver, AAS standard solution, Specpure™ Ag 1000μg/mL, Thermo Scientific Chemicals
Silver, AAS standard solution, Specpure™ Ag 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silver, AAS standard solution, Specpure™ Ag 1000μg/mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF088096.AE100 mL
Catalog number ALF088096.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver, AAS standard solution
Name NoteAg 1000μg/mL
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c