Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Thermo Scientific Chemicals
Gold, AAS standard solution, Specpure™ Au 1000μg/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088068.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialGold, AAS standard solution
Name NoteAu 1000μg/mL
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Gold, AAS standard solution is used for the determination of gold by flame atomic absorption spectrometry (FAAS) and graphite furnace atomic absorption spectrometry (GFAAS).