Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL
Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL
Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL
Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL
Thermo Scientific Chemicals
Cobalt, AAS standard solution, Specpure™ Co 1000μg/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088060.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt, AAS standard solution
Name NoteCo 1000μg/mL
Health Hazard 1H290-H314-H335-H350i-H360F
Health Hazard 2GHS H Statement
H334-H314-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c